Skip to Content
Merck
All Photos(2)

Documents

523259

Sigma-Aldrich

α-Methyl-9-anthracenemethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O
CAS Number:
Molecular Weight:
222.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

124-128 °C (lit.)

SMILES string

CC(O)c1c2ccccc2cc3ccccc13

InChI

1S/C16H14O/c1-11(17)16-14-8-4-2-6-12(14)10-13-7-3-5-9-15(13)16/h2-11,17H,1H3

InChI key

WRADANNQOTZBDC-UHFFFAOYSA-N

General description

α-Methyl-9-anthracenemethanol is an aromatic alcohol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ashtari et al.
Journal of chromatography. A, 1409, 89-107 (2015-07-30)
Molecular dynamics simulations are employed to examine the selectivity of four proline-based chiral stationary phases in two solvent environments, a relatively apolar n-hexane/2-propanol solvent and a polar water/methanol solvent. The four chiral surfaces are based on a BOC-terminated diproline, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service