Skip to Content
Merck
All Photos(1)

Key Documents

521086

Sigma-Aldrich

1-Butyl-4-ethynylbenzene

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$84.40

$84.40


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$84.40

About This Item

Linear Formula:
CH3(CH2)3C6H4C≡CH
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.40


Estimated to ship onMay 24, 2025


Request a Bulk Order

Assay

95%

refractive index

n20/D 1.527 (lit.)

density

0.906 g/mL at 25 °C (lit.)

SMILES string

CCCCc1ccc(cc1)C#C

InChI

1S/C12H14/c1-3-5-6-12-9-7-11(4-2)8-10-12/h2,7-10H,3,5-6H2,1H3

InChI key

ZVWWYEHVIRMJIE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadezhda Romanchikova et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 765-772 (2013-11-20)
A novel matrix metalloproteinase-2 (MMP-2) inhibitor JaZ-30, which belongs to the class of C(2)-monosubstituted aziridine - aryl-1,2,3-triazole conjugates, was developed. MTT and crystal violet assays were used to determine cytotoxicity- IC(50) values of compound JaZ-30 on melanoma cell line B16
Synthesis and characterization of the multi-photon absorption and excited-state properties of a neat liquid 4-propyl 4'-butyl diphenyl acetylene
Khoo LC, et al.
Journal of Materials Chemistry, 19(40), 7525-7525 (2009)
Regioselective One-Pot Synthesis of Isocoumarins and Phthalides from 2-Iodobenzoic Acids and Alkynes by Temperature Control.
Kumar MR, et al.
Advanced Synthesis & Catalysis, 355(16), 3221-3230 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service