Skip to Content
Merck
All Photos(1)

Documents

520934

Sigma-Aldrich

Cesium orthovanadate

≥99.9% trace metals basis (purity excludes 0.5-1% aluminum)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cs3VO4
CAS Number:
Molecular Weight:
513.66
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.9% trace metals basis (purity excludes 0.5-1% aluminum)

form

powder

SMILES string

[Cs+].[Cs+].[Cs+].[O-][V]([O-])([O-])=O

InChI

1S/3Cs.4O.V/q3*+1;;3*-1;

InChI key

CZUUFJSJUCXOQF-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E L Leung et al.
British journal of cancer, 98(11), 1803-1809 (2008-05-29)
The present study determines if (1) basal protein levels of nitric oxide (NO) synthases (eNOS, iNOS, and nNOS) are different in cisplatin-sensitive (OV2008) and counterpart cisplatin-resistant (C13(*)) human ovarian cancer cells, (2) cisplatin alters NOS levels, (3) NO donor causes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service