Skip to Content
Merck
All Photos(2)

Key Documents

520144

Sigma-Aldrich

Calcium thiocyanate tetrahydrate

95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$117.00

$117.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$117.00

About This Item

Linear Formula:
Ca(SCN)2·4H2O
CAS Number:
Molecular Weight:
228.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

$117.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

for analytical purposes

Assay

95%

form

solid

mp

42.5-46.5 °C (lit.)

SMILES string

O.O.O.O.N#CS[Ca]SC#N

InChI

1S/2CHNS.Ca.4H2O/c2*2-1-3;;;;;/h2*3H;;4*1H2/q;;+2;;;;/p-2

InChI key

HOQNOUSJUGLELS-UHFFFAOYSA-L

Application

  • Roar Myrdal: Discusses the use of calcium thiocyanate along with sodium thiocyanate and calcium nitrate in concrete as accelerating admixtures, highlighting their effects on the setting time and strength development (H Justnes).
  • Gas permeability of cellulose aerogels with a designed dual pore space system: Investigates the role of calcium thiocyanate tetrahydrate in the creation of cellulose aerogels, focusing on its impact on the structural and physical properties of the aerogels, crucial for applications such as insulation and filtration (K Ganesan et al., 2019).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myung-Joon Jeong et al.
Polymers, 11(9) (2019-09-25)
The dissolution process of cellulose aerogels is an important part of their production. However, if the cellulose is severely degraded during the dissolution process, the quality may be low. To evaluate the degradation of cellulose during the dissolution process using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service