All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H5NOS
CAS Number:
Molecular Weight:
127.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
98%
mp
181-184 °C (lit.)
SMILES string
NC(=O)c1ccsc1
InChI
1S/C5H5NOS/c6-5(7)4-1-2-8-3-4/h1-3H,(H2,6,7)
InChI key
DAUYIKBTMNZABP-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Juan C Vidal et al.
Mikrochimica acta, 187(3), 183-183 (2020-02-24)
New electrochemical sensors are described for the rapid quantification of silver nanoparticles (AgNPs). They are based on the immobilization of L-cysteine on a glassy carbon electrode (GCE) and on the formation of electropolymerized oligomers on the GCE. Ligands with amino
Amar Hekmat Mahdi et al.
Cancer research, 78(14), 3969-3981 (2018-05-10)
The BRCA1-PALB2-BRCA2 axis plays an essential role in DNA homologous recombination repair, defect in which drives genome instability and cancer development. How cells with defects in this pathway respond to DNA damage in vivo and how tumors develop from these
Jordi Codony-Servat et al.
Clinical lung cancer, 20(3), 167-177 (2019-03-20)
Epidermal growth factor receptor (EGFR) pathway deregulation promotes the acquisition of stemlike properties in non-small-cell lung cancer. EGFR inhibition through NOTCH enriches lung cancer stem cells (CSCs). Src through Yes-associated protein 1 (YAP1) activates NOTCH. Signal transduction and activator of
Katrin Schweitzer et al.
Journal of cellular and molecular medicine, 20(1), 58-70 (2015-10-16)
Cullin-RING-ubiquitin-ligase (CRL)-dependent ubiquitination of the nuclear factor kappa B (NF-κB) inhibitor IκBα and its subsequent degradation by the proteasome usually precede NF-κB/RelA nuclear activity. Through removal of the CRL-activating modification of their cullin subunit with the ubiquitin (Ub)-like modifier NEDD8
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service