Skip to Content
Merck
All Photos(1)

Key Documents

518719

Sigma-Aldrich

4-Bromo-3-fluoroaniline

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$39.20

$39.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$39.20

About This Item

Linear Formula:
BrC6H3(F)NH2
CAS Number:
Molecular Weight:
190.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$39.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.471 (lit.)

bp

65-68 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Nc1ccc(Br)c(F)c1

InChI

1S/C6H5BrFN/c7-5-2-1-4(9)3-6(5)8/h1-3H,9H2

InChI key

YTMVYYAKOPIJCZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorinated isatins and some of their heterocyclic derivatives.
Yen VQ, et al.
The Journal of Organic Chemistry, 23(12), 1858-1861 (1958)
Gang Li et al.
Chemical & pharmaceutical bulletin, 63(2), 143-146 (2014-12-11)
An improved and efficient method for the preparation of torezolid based on Suzuki cross-coupling reaction as the key step was developed on a gram scale in five steps. The total yield was 44% and the optical purity of torezolid by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service