Skip to Content
Merck
All Photos(2)

Key Documents

516112

Sigma-Aldrich

Cyclopropylacetonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$118.00

$118.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$118.00

About This Item

Linear Formula:
C3H5CH2CN
CAS Number:
Molecular Weight:
81.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$118.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.4235 (lit.)

bp

142-144 °C (lit.)

density

0.878 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

N#CCC1CC1

InChI

1S/C5H7N/c6-4-3-5-1-2-5/h5H,1-3H2

InChI key

FAUQRRGKJKMEIW-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.0 °F - closed cup

Flash Point(C)

43.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Synthesis of Methyl Cyclopropylacetate by Palladium (II) Acetate Catalysed Reaction Of Diazomethane with Vinyl Group.
Basnak I, et al.
Synthetic Communications, 22(5), 773-782 (1992)
Phyllis A Leber et al.
Molecules (Basel, Switzerland), 19(2), 1527-1543 (2014-01-30)
The title compound 1-exo (with minor amounts of its C8 epimer 1-endo) was prepared by Wolff-Kishner reduction of the cycloadduct of 1,3-cyclohexadiene and cyclopropylketene. The [1,3]-migration product 2-endo was synthesized by efficient selective cyclopropanation of endo-5-vinylbicyclo[2.2.2]oct-2-ene at the exocyclic π-bond.
Charles E Mowbray et al.
Journal of medicinal chemistry, 58(24), 9615-9624 (2015-11-17)
Visceral leishmaniasis is a severe parasitic disease that is one of the most neglected tropical diseases. Treatment options are limited, and there is an urgent need for new therapeutic agents. Following an HTS campaign and hit optimization, a novel series
Jonathan D Rosen et al.
Tetrahedron letters, 50(7), 785-789 (2010-02-18)
An optimized total synthesis of the 2-amino-6-chloro-4-cyclopropyl-7-fluoro-5-methoxy-pyrido[1,2-c]pyrimidine-1,3-dione core structure of a new fluoroquinolone-like class of antibacterial agents is described. This synthesis is highlighted by a nearly quantitative ring-closing reaction to form the pyrido[1,2-c]pyrimidine core. This bicyclic ring system serves as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service