514586
4-Methoxy-TEMPO
97%
Synonym(s):
4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H20NO2
CAS Number:
Molecular Weight:
186.27
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
40.5-44 °C (lit.)
storage temp.
2-8°C
SMILES string
COC1CC(C)(C)N([O])C(C)(C)C1
InChI
1S/C10H20NO2/c1-9(2)6-8(13-5)7-10(3,4)11(9)12/h8H,6-7H2,1-5H3
InChI key
SFXHWRCRQNGVLJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-Methoxy-TEMPO (4-methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy) is an aminoxyl free radical commonly employed for the oxidation reaction of alcohols (primary/secondary). It has been proposed as hindered amine light stabiliser (HALS) model compound. Its reaction with irradiated dilute aqueous suspensions of photocatalytic nanoparticulate titanium dioxide was investigated by electron spin resonance (ESR) and electrospray ionization mass-spectrometry (ESI-MS).
Application
4-Methoxy-TEMPO may be employed as catalyst for the synthesis of 2-substituted benzoxazoles, via aerobic oxidation reaction.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
David L Marshall et al.
Organic & biomolecular chemistry, 9(13), 4936-4947 (2011-05-21)
Potenital pathways for the deactivation of hindered amine light stabilisers (HALS) have been investigated by observing reactions of model compounds--based on 4-substituted derivatives of 2,2,6,6-tetramethylpiperidine-N-oxyl (TEMPO)--with hydroxyl radicals. In these reactions, dilute aqueous suspensions of photocatalytic nanoparticulate titanium dioxide were
Xican Li et al.
Molecules (Basel, Switzerland), 24(11) (2019-05-31)
To probe the effect of 3',8″-dimerization on antioxidant flavonoids, acacetin and its 3',8″-dimer isoginkgetin were comparatively analyzed using three antioxidant assays, namely, the ·O2- scavenging assay, the Cu2+ reducing assay, and the 2,2'-azino bis(3-ethylbenzothiazolin-6-sulfonic acid) radical scavenging assay. In these
Xiaoqing Guo et al.
Toxicological sciences : an official journal of the Society of Toxicology, 163(1), 214-225 (2018-02-01)
TEMPO (2, 2, 6, 6-tetramethylphiperidine-1-oxyl) and its derivatives are stable free radical nitroxides widely used in the field of chemistry, biology, and pharmacology. TEMPO was previously found to be mutagenic and to induce micronuclei in mammalian cells. In this study
D Metodiewa et al.
Anticancer research, 19(6B), 5259-5264 (2000-03-04)
A stable nitroxide radical named Metexyl (4-methoxy-2,2,6,6-tetramethylpiperidine-1-oxyl) was synthesized and its antioxidant and antitumor properties were investigated and compared with these of another nitroxide derivatives previously designed in our laboratories. Three experimental models were used: xanthine/xanthine oxidase system, pulse radiolysis
Kevin M Church et al.
Nucleosides, nucleotides & nucleic acids, 23(11), 1723-1738 (2004-12-16)
Our work outlines the use of oxoammonium salts in a formal 1,2 addition process to olefins giving nucleoside analogs as products. Specifically, oxoammonium salts can be added to a solution of olefin and silylated heterocycle to give Methoxy TEMPO substituted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service