Skip to Content
Merck
All Photos(1)

Key Documents

514381

Sigma-Aldrich

Titanium(III) chloride

≥99.995% trace metals basis

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.995% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: titanium

mp

440 °C (dec.) (lit.)

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used for the vapor-phase formation of intermetallic compounds with ultrafine particle size.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gnana S Siluvai et al.
Biochemistry, 48(51), 12133-12144 (2009-11-20)
Sco-like proteins contain copper bound by two cysteines and a histidine residue. Although their function is still incompletely understood, there is a clear involvement with the assembly of cytochrome oxidases that contain the Cu(A) center in subunit 2, possibly mediating
Catalytic oxidation of hydrazo derivatives promoted by a TiCl3/HBr system.
Eyal Drug et al.
Journal of the American Chemical Society, 129(45), 13784-13785 (2007-10-26)
D J Kushner et al.
Analytical biochemistry, 133(1), 116-119 (1983-08-01)
Hydroxamic acid siderophores were observed to be inactivated by exposure to titanium(III) chloride. To study the reaction, a series of eight model hydroxamic acids were prepared and reacted with titanium(III) chloride. The products were shown by ir and NMR comparisons
C Noubactep et al.
Journal of hazardous materials, 132(2-3), 202-212 (2005-11-08)
The effectiveness of elemental iron (Fe(0)) to remove uranium (U) from the aqueous phase has been demonstrated. While the mitigation effect is sure, discrepancies in the removal mechanism have been reported. The objective of this study was to investigate the
Investigation into the effectiveness of surface treatment on poly(methyl methacrylate) when exposed in the mouth.
M D Murray
The Journal of prosthetic dentistry, 59(3), 368-373 (1988-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service