Skip to Content
Merck
All Photos(3)

Key Documents

513164

Sigma-Aldrich

Iodopyrazine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$65.70

$65.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$65.70

About This Item

Empirical Formula (Hill Notation):
C4H3IN2
CAS Number:
Molecular Weight:
205.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$65.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.655 (lit.)

bp

109-110 °C/34 mmHg (lit.)

density

2.086 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

Ic1cnccn1

InChI

1S/C4H3IN2/c5-4-3-6-1-2-7-4/h1-3H

InChI key

OYWPFIUVDKHHGQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron (II) isothiocyanate complexes with substituted pyrazines: Experimental and theoretical views on their electronic structure.
Shylin SI, et al.
Polyhedron, 87, 147-155 (2015)
The Synthesis and Properties of Iodopyrazines.
Hirschberg A and Spoerri PE.
The Journal of Organic Chemistry, 26(6), 1907-1912 (1961)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service