512931
3,4-(Methylenedioxy)phenylmagnesium bromide solution
1.0 M in THF: toluene (1:1)
About This Item
Recommended Products
reaction suitability
reaction type: Grignard Reaction
concentration
1.0 M in THF: toluene (1:1)
density
1.04 g/mL at 25 °C
SMILES string
Br[Mg]c1ccc2OCOc2c1
InChI
1S/C7H5O2.BrH.Mg/c1-2-4-7-6(3-1)8-5-9-7;;/h1,3-4H,5H2;1H;/q;;+1/p-1
InChI key
JFUHZUGUHBLMLB-UHFFFAOYSA-M
Application
Signal Word
Danger
Hazard Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
14.0 °F - closed cup
Flash Point(C)
-10 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service