Skip to Content
Merck
All Photos(1)

Key Documents

511862

Sigma-Aldrich

9H-Fluoren-2-yl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO
CAS Number:
Molecular Weight:
207.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

69-71 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=Nc1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C14H9NO/c16-9-15-12-5-6-14-11(8-12)7-10-3-1-2-4-13(10)14/h1-6,8H,7H2

InChI key

HJROCLJFUOYQNP-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose A Castro-Osma et al.
Dalton transactions (Cambridge, England : 2003), 44(27), 12388-12400 (2014-12-24)
A series of alkyl organoaluminium complexes based on bulky heteroscorpionate ligands were designed as catalysts for the ring-opening polymerisation of cyclic esters. Thus, the treatment of AlX3 (X = Me, Et) with bulky acetamide or thioacetamide heteroscorpionate ligands nbptamH (1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service