Skip to Content
Merck
All Photos(2)

Key Documents

499986

Sigma-Aldrich

4-Dibenzothienylboronic acid

≥95.0%

Synonym(s):

4-Dibenzothiopheneboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$46.20
5 G
$50.80

$46.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$46.20
5 G
$50.80

About This Item

Empirical Formula (Hill Notation):
C12H9BO2S
CAS Number:
Molecular Weight:
228.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$46.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0%

mp

327-330 °C (lit.)

SMILES string

OB(O)c1cccc2c3ccccc3sc12

InChI

1S/C12H9BO2S/c14-13(15)10-6-3-5-9-8-4-1-2-7-11(8)16-12(9)10/h1-7,14-15H

InChI key

GOXNHPQCCUVWRO-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service