Skip to Content
Merck
All Photos(3)

Key Documents

499811

Sigma-Aldrich

Lithium

granular, 99% trace metals basis

Synonym(s):

Lithium atom, Lithium element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li
CAS Number:
Molecular Weight:
6.94
EC Number:
MDL number:
UNSPSC Code:
12141803
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99% trace metals basis

form

granular

reaction suitability

reagent type: reductant

resistivity

9.446 μΩ-cm, 20°C

bp

1342 °C (lit.)

mp

180 °C (lit.)

density

0.534 g/mL at 25 °C (lit.)

SMILES string

[Li]

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Granular lithium is a form of lithium metal that has been ground into small granules. Lithium is a soft, silver-white metal that is highly reactive and flammable. Lithium has a low density of 0.534 g/cm3, which makes it one of the lightest metals. Like the other alkali metals, lithium has a low melting point (180 °C) and is a good conductor of electricity and heat. Granular lithium is highly reactive and behaves as a strong reducing agent. It must be handled with caution due to its potential to ignite or explode. Lithium reacts readily with water and oxygen to form lithium hydroxide and lithium oxide, respectively, and even reacts with nitrogen gas at room temperature, forming lithium nitride. Typically, lithium is stored in an argon glovebox and airtight containers to prevent exposure to moisture, air, and nitrogen. To make granular lithium, the metal is first extracted from ores such as spodumene or lepidolite using a process called lithium carbonate conversion. After the lithium has been purified to a high degree of purity, it is milled into granules by rotating blades.
Lithium is a low-density alkali metal that is widely used as an anode material in rechargeable and non-rechargeable batteries. It is also used to alloy with aluminum and magnesium to make them stronger and lighter.

Application

Lithium can be used as:
  • A precursor to synthesize Li-based alloys such as LiAl and LiSi alloys, which are applicable as anode materials in the field of energy conversion and storage.[1]
  • A reducing agent in the reduction of zirconium oxide compounds in molten LiCl salt.[2]
  • As starting material to synthesize a reducing agent(1,4-bis(trimethylgermyl)-1.4-dihydropyrazine), for the fabrication of nickel metal films.[3]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A low-temperature thermal ALD process for nickel utilizing dichlorobis(triethylphosphine)nickel(ii) and 1{,}4-bis(trimethylgermyl)-1{,}4-dihydropyrazine"
Anton Vihervaara,
Dalton Transactions, 51, 10898-10908 (2022)
Anode performance of lithium--silicon alloy prepared by mechanical alloying for use in all-solid-state lithium secondary batteries
Hye Won Park, et al.
Japanese Journal of Applied Physics, 53, 08NK02-08NK02 (2014)
Reduction of zirconium oxide compounds by lithium metal as a reductant in molten LiCl salt
Eun-Young Choi and Dong Hyun Heo
Journal of Nuclear Materials, 512, 193-198 (2018)
Sureshbabu Guduguntla et al.
The Journal of organic chemistry, 78(17), 8274-8280 (2013-08-22)
An efficient one-pot synthesis of optically active β-alkyl-substituted alcohols through a tandem copper-catalyzed asymmetric allylic alkylation (AAA) with organolithium reagents and reductive ozonolysis is presented. Furthermore, hydroboration-oxidation following the Cu-catalyzed AAA leads to the corresponding homochiral γ-alkyl-substituted alcohols.
Jian Jiang et al.
Nanoscale, 5(17), 8105-8113 (2013-07-26)
Controlled integration of multiple semiconducting oxides into each single unit of ordered nanotube arrays is highly desired in scientific research for the realization of more attractive applications. We herein report a diffusion-controlled solid-solid route to evolve simplex Co(CO3)0.5(OH)0.11H2O@TiO2 core-shell nanowire

Questions

1–6 of 6 Questions  
  1. Are there trace amounts of mineral oil on this lithium? How is it handelled prior to being bottled under argon? After production, is argon the only inert atmosphere used?

    1 answer
    1. For this product, argon gas is the only inert atmosphere used. Unfortunately, other information on the method of preparation of this product, such as whether or not mineral oil is used, is held as proprietary by the original manufacturer.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. This lithium is stated to be granular. What is the average particle diameter of the granules?

    1 answer
    1. The particle size is not determined for this product.

      Helpful?

  5. What atmosphere is the granular lithium (product 499811-25G) shipped under? Specifically, is it under an inert atmosphere in the bottle, or under a natural atmosphere?

    1 answer
    1. The granular lithium (product 499811-25G) is packaged under argon, which provides an inert atmosphere within the bottle.

      Helpful?

  6. Greeting! Where could i found expiration date?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service