Skip to Content
Merck
All Photos(1)

Key Documents

497762

Sigma-Aldrich

4-Chlorobenzylzinc chloride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$413.00

$413.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$413.00

About This Item

Linear Formula:
ClC6H4CH2ZnCl
CAS Number:
Molecular Weight:
226.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$413.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Quality Level

concentration

0.5 M in THF

density

0.96 g/mL at 25 °C

functional group

chloro

storage temp.

2-8°C

SMILES string

Cl[Zn]Cc1ccc(Cl)cc1

InChI

1S/C7H6Cl.ClH.Zn/c1-6-2-4-7(8)5-3-6;;/h2-5H,1H2;1H;/q;;+1/p-1

InChI key

JIWPVHAJWTWEJB-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-6.2 °F

Flash Point(C)

-21.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service