Skip to Content
Merck
All Photos(1)

Key Documents

497231

Sigma-Aldrich

5-Methoxy-4-methylindole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO
CAS Number:
Molecular Weight:
161.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

76-79 °C (lit.)

SMILES string

COc1ccc2[nH]ccc2c1C

InChI

1S/C10H11NO/c1-7-8-5-6-11-9(8)3-4-10(7)12-2/h3-6,11H,1-2H3

InChI key

RISMXZVKSIWLMK-UHFFFAOYSA-N

General description

5-Methoxy-4-methylindole or 5-methoxy-4-methyl-1H-indole is a substituted 1H-indole. Its efficacy as a substrate for indigoid production has been evaluated.

Application

  • Reactant for preparation of β-carboline-1-carboxylic acids as inhibitors of mitogen activated protein kinase-activated protein kinase 2
  • Reactant for preparation of indolyl and isoquinolinyl anthranilates as PPARδ partial agonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongliu-Liu Wu et al.
Chemistry & biodiversity, 2(1), 51-65 (2006-12-29)
Glycogen synthase kinase-3 (GSK-3) is a potential drug target for a number of human diseases. Some indigoids have been found to be potent inhibitors of GSK-3, and individual compounds with better activity, specificity, and solubility are desired. In this work

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service