Skip to Content
Merck
All Photos(1)

Key Documents

496707

Sigma-Aldrich

Poly(vinylidene chloride-co-acrylonitrile)

average Mn ~80,000, average Mw ~150,000

Synonym(s):

Acrylonitrile-vinylidene chloride polymer, Acrylonitrile-vinylidene copolymer, Poly(vinylidene chloride-acrylonitrile), Poly(vinylidene chloride-acrylonitrile) copolymer, Polyacrylonitrile-vinylidene chloride copolymer, Vinylidene chloride-acrylonitrile polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CCl2)x[CH2CH(CN)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

mol wt

average Mn ~80,000
average Mw ~150,000

transition temp

Tg 50 °C

density

1.6 g/mL at 25 °C

SMILES string

ClC(Cl)=C.C=CC#N

InChI

1S/C3H3N.C2H2Cl2/c1-2-3-4;1-2(3)4/h2H,1H2;1H2

InChI key

QLUXVUVEVXYICG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan Schreml et al.
Experimental dermatology, 20(7), 550-554 (2011-03-30)
In cutaneous wound healing, the role of oxygen in vivo is poorly understood. We studied wound surface pO(2) during physiological wound healing in humans. Split-thickness skin graft donor sites (n=12) served as standardized wound models. Wound surface pO(2) was measured
L Litauszki et al.
Journal of biomedical materials research, 35(1), 1-8 (1997-04-01)
Poly(ethylene oxide) (PEO) was immobilized by the Williamson ether synthesis onto halogenated surfaces such as poly(vinylidene chloride), surface-brominated polypropylene, and surface-brominated poly(ethylene terephthalate). PEO (Mw 20,000) was converted to its sodium salt and reacted for various times with the halogen-containing
S Baumgart
Pediatrics, 74(6), 1022-1028 (1984-12-01)
Eight very low-birth-weight premature infants (mean birth weight 1.11 +/- 0.05 [SEM]kg, mean gestation 30 +/- 1 weeks, and mean age 9 +/- 2 days) were studied under servocontrolled radiant warmers with and without a loosely fitted, transparent, and flexible
R C Strohman et al.
In vitro cellular & developmental biology : journal of the Tissue Culture Association, 26(2), 201-208 (1990-02-01)
Primary muscle cell cultures consisting of single myocytes and fibroblasts are grown on flexible, optically clear biomembranes. Muscle cell growth, fusion and terminal differentiation are normal. A most effective membrane for these cultures is commercially available Saran Wrap. Muscle cultures
Nirav K Patel et al.
British journal of nursing (Mark Allen Publishing), 16(18), 1140-1142 (2007-12-13)
Clubfoot or congenital talipes equinovarus is a common condition affecting babies and non-surgical treatment involves serial manipulation and plaster casting for many weeks. The casts are susceptible to soiling during this time, which makes management and child care even more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service