Skip to Content
Merck
All Photos(2)

Key Documents

493449

Sigma-Aldrich

2-Benzofurancarboxaldehyde

97%

Synonym(s):

1-Benzofuran-2-carbaldehyde, 2-Benzofurancarbaldehyde, 2-Formylbenzofuran, Benzo[b]-2-furfural, Benzo[b]furan-2-aldehyde, Benzo[b]furan-2-carboxaldehyde, Coumarilaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$112.00
5 G
$283.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$112.00
5 G
$283.00

About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.633 (lit.)

bp

135 °C/18 mmHg (lit.)

density

1.206 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

O=Cc1cc2ccccc2o1

InChI

1S/C9H6O2/c10-6-8-5-7-3-1-2-4-9(7)11-8/h1-6H

InChI key

ADDZHRRCUWNSCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Benzofurancarboxaldehyde, also known as benzo[b]-2-furfural, is an α,β-unsaturated aldehyde.

Application

2-Benzofurancarboxaldehyde may be used in the preparation of (E)-3-(benzofuran-2′-ylmethylidene)-1-methyl-2-indolinone[1] and 1-(benzofuran-2-yl)methanol.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrogenations without hydrogen: titania photocatalyzed reductions of maleimides and aldehydes.
Manley DW, et al.
Molecules (Basel), 19(9), 15324-15338 (2014)
Telescoped Enolate Arylation/HWE Procedure for the Preparation of 3-Alkenyl-Oxindoles: The First Synthesis of Soulieotine.
Millemaggi A, et al.
European Journal of Organic Chemistry, 2009(18), 2947-2952 (2009)
Niina Tani et al.
Bioorganic & medicinal chemistry, 22(23), 6655-6664 (2014-12-03)
Inhibition of CYP2A6-mediated nicotine metabolism can reduce cigarette smoking. We sought potent and selective CYP2A6 inhibitors to be used as leads for drugs useful in smoking reduction therapy, by evaluating CYP2A6 inhibitory effect of novel formyl, alkyl amine or carbonitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service