Skip to Content
Merck
All Photos(1)

Documents

493236

Sigma-Aldrich

Phenylacetic-d7 acid

98 atom % D

Synonym(s):

Benzene-d5-acetic-d2 acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD2CO2H
CAS Number:
Molecular Weight:
143.19
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

bp

265 °C (lit.)

mp

77-79 °C (lit.)

density

1.136 g/mL at 25 °C

mass shift

M+7

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C(O)=O

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)/i1D,2D,3D,4D,5D,6D2

InChI key

WLJVXDMOQOGPHL-XZJKGWKKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

269.6 °F

Flash Point(C)

132.00 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service