490407
Chloropentamethyldisilane
97%
Synonym(s):
1-Chloro-1,1,2,2,2-pentamethyldisilane, Chlorodimethyl(trimethylsilyl)silane, Chloropentamethyldisilane, Pentamethylchlorodisilane
About This Item
Assay
97%
form
liquid
refractive index
n20/D 1.4415 (lit.)
bp
134-136 °C (lit.)
density
0.862 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)(C)[Si](C)(C)Cl
InChI
1S/C5H15ClSi2/c1-7(2,3)8(4,5)6/h1-5H3
InChI key
GJCAUTWJWBFMFU-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
102.2 °F - closed cup
Flash Point(C)
39 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer
Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service