Skip to Content
Merck
All Photos(1)

Key Documents

488518

Sigma-Aldrich

Chloroacetic acid-13C2

99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$2,340.00

$2,340.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

Select a Size

Change View
250 MG
$2,340.00

About This Item

Linear Formula:
Cl13CH213CO2H
CAS Number:
Molecular Weight:
96.48
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$2,340.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

isotopic purity

99 atom % 13C

form

solid

bp

189 °C (lit.)

mp

60-63 °C (lit.)

density

1.613 g/mL

mass shift

M+2

SMILES string

O[13C](=O)[13CH2]Cl

InChI

1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)/i1+1,2+1

InChI key

FOCAUTSVDIKZOP-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service