Skip to Content
Merck
All Photos(1)

Key Documents

488402

Sigma-Aldrich

Calcium nitrate-15N2

10 atom % 15N

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$1,160.00

$1,160.00


Estimated to ship onMarch 19, 2025


Request more information

Select a Size

Change View
25 G
$1,160.00

About This Item

Linear Formula:
Ca(15NO3)2
CAS Number:
Molecular Weight:
166.07
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

$1,160.00


Estimated to ship onMarch 19, 2025


Request more information

isotopic purity

10 atom % 15N

Quality Level

form

solid

mol wt

164.28 by atom % calculation (10 atom % 15N)

mp

561 °C (lit.)

application(s)

agriculture

mass shift

M+2

SMILES string

[Ca++].[O-][15N+]([O-])=O.[O-][15N+]([O-])=O

InChI

1S/Ca.2NO3/c;2*2-1(3)4/q+2;2*-1/i;2*1+1

InChI key

ZCCIPPOKBCJFDN-IMSGLENWSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Tracer techniques using isotopic N have been employed since the 1940s to investigate such topics as N transformations and cycling in agricultural and other soils, N sources for crop uptake, the fate and behavior of fertilizer N applied to soils, the nature and extent of soil and fertilizer losses, and fertilizer N uptake efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service