Skip to Content
Merck
All Photos(1)

Key Documents

487740

Sigma-Aldrich

4-Oxo-TEMPO-d16,15N, free radical

98 atom % D, 98 atom % 15N, for ESR-spectroscopy

Synonym(s):

4-Oxo-2,2,6,6-tetramethylpiperidine-d16,1-15N-1-oxyl, TEMPONE-d16,1-15N

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$2,050.00

$2,050.00


Estimated to ship onMay 27, 2025


Request more information

Select a Size

Change View
100 MG
$2,050.00

About This Item

Empirical Formula (Hill Notation):
C9D1615NO2
CAS Number:
Molecular Weight:
187.32
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$2,050.00


Estimated to ship onMay 27, 2025


Request more information

grade

for ESR-spectroscopy

isotopic purity

98 atom % 15N
98 atom % D

Assay

99% (CP)

form

solid

mp

35 °C (lit.)

mass shift

M+17

SMILES string

[2H]C([2H])([2H])C1([15N]([O])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C(=O)C1([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C9H16NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,10+1

InChI key

WSGDRFHJFJRSFY-ZDYDSYIGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service