Assay
95%
refractive index
n20/D 1.568 (lit.)
bp
282 °C (lit.)
density
1.375 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
Clc1ccc(CN=C=O)cc1Cl
InChI
1S/C8H5Cl2NO/c9-7-2-1-6(3-8(7)10)4-11-5-12/h1-3H,4H2
InChI key
WLNAHQZFPSSVTP-UHFFFAOYSA-N
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service