Skip to Content
Merck
All Photos(1)

Key Documents

485535

Sigma-Aldrich

DL-Dithiothreitol-d10

98 atom % D, 98% (CP)

Synonym(s):

Cleland’s reagent-d10

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$3,560.00

$3,560.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

Select a Size

Change View
500 MG
$3,560.00

About This Item

Linear Formula:
DSCD2CD(OD)CD(OD)CD2SD
CAS Number:
Molecular Weight:
164.31
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NA.12

$3,560.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

technique(s)

NMR: suitable
bio NMR: suitable

mp

42-44 °C (lit.)

mass shift

M+10

storage temp.

2-8°C

SMILES string

[2H]OC([2H])(C([2H])([2H])S[2H])C([2H])(O[2H])C([2H])([2H])S[2H]

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/i1D2,2D2,3D,4D,5D,6D/hD2

InChI key

VHJLVAABSRFDPM-BSSDICLPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service