Skip to Content
Merck
All Photos(1)

Key Documents

485071

Sigma-Aldrich

3′,6′-Dichlorofluoran

95%

Synonym(s):

Fluorescein dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Cl2O3
CAS Number:
Molecular Weight:
369.20
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

95%

mp

253-255 °C (lit.)

SMILES string

Clc1ccc2c(Oc3cc(Cl)ccc3C24OC(=O)c5ccccc45)c1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service