Skip to Content
Merck
All Photos(1)

Key Documents

483575

Sigma-Aldrich

Diamond

monocrystalline powder, ~1 μm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12352103

form

monocrystalline powder

particle size

~1 μm

density

3.5 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

Looking for similar products? Visit Product Comparison Guide

General description

Mono- and polycrystalline diamond can be prepared by high pressure high temperature (HPHT) and chemical vapor deposition (CVD).[1]

Application

Monocrystalline diamond paste based electrodes may be used as electrochemical sensors for the determination of Fe (II) in pharmaceutical products.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave plasma deposition and mechanical treatment of single crystals and polycrystalline diamond films
Ashkihazi EE, et al.
Inorganic Materials, 5(3), 230-236 (2014)
Monocrystalline Diamond Paste-Based Electrodes and Their Applications for the Determination of Fe(II) in Vitamin
Stefan RI and Bairu SG
Analytical Chemistry, 75(20), 5394-5398 (2003)
Sinead P Blaber et al.
PloS one, 8(1), e52997-e52997 (2013-01-10)
Stem cells are increasingly the focus of translational research as well as having emerging roles in human cellular therapy. To support these uses there is a need for improved methods for in vivo cell localization and tracking. In this study
D Le Sage et al.
Nature, 496(7446), 486-489 (2013-04-27)
Magnetic imaging is a powerful tool for probing biological and physical systems. However, existing techniques either have poor spatial resolution compared to optical microscopy and are hence not generally applicable to imaging of sub-cellular structure (for example, magnetic resonance imaging)
Alexandre Barras et al.
Nanoscale, 5(6), 2307-2316 (2013-02-12)
Bacterial attachment and subsequent biofilm formation on biotic surfaces or medical devices is an increasing source of infections in clinical settings. A large proportion of these biofilm-related infections are caused by Escherichia coli, a major nosocomial pathogen, in which the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service