Skip to Content
Merck
All Photos(2)

Key Documents

483354

Sigma-Aldrich

Scandium(III) triflate

99.995% trace metals basis

Synonym(s):

Sc(OTf)3, Scandium tris(trifluoromethanesulfonate), Scandium(III) trifluoromethanesulfonate, Trifluoromethanesulfonic acid scandium(III) salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$110.00

$110.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$110.00

About This Item

Linear Formula:
Sc(SO3CF3)3
CAS Number:
Molecular Weight:
492.16
Beilstein:
8510151
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$110.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

99.995% trace metals basis

reaction suitability

core: scandium
reagent type: catalyst

mp

>300 °C

SMILES string

[Sc+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Sc/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

HZXJVDYQRYYYOR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholho Choe et al.
Dalton transactions (Cambridge, England : 2003), 44(19), 9182-9192 (2015-04-24)
Redox-inactive metal ions can modulate the reactivity of redox-active metal ions in a variety of biological and chemical oxidations. Many synthetic models have been developed to help address the elusive roles of these redox-inactive metal ions. Using a non-heme manganese(II)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service