Skip to Content
Merck
All Photos(1)

Key Documents

481386

SAFC

Epichlorohydrin

≥99%

Synonym(s):

(±)-Epichlorohydrin, (±)-2-(Chloromethyl)oxirane, 1-Chloro-2,3-epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
Beilstein:
79785
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

3.2 (vs air)

vapor pressure

13.8 mmHg ( 21.1 °C)

Assay

≥99%

autoignition temp.

779 °F

expl. lim.

21 %

impurities

≤0.1% water

refractive index

n20/D 1.438 (lit.)

bp

115-117 °C (lit.)

mp

−57 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

SMILES string

ClCC1CO1

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2

InChI key

BRLQWZUYTZBJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F

Flash Point(C)

28 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An evaluation of the genetic toxicity of epichlorohydrin. A report of an expert group of the International Commission for Protection against Environmental Mutagens and Carcinogens.
R J Srám et al.
Mutation research, 87(3), 299-319 (1981-11-01)
Swetha Inturi et al.
ACS nano, 9(11), 10758-10768 (2015-10-22)
Notwithstanding rapid advances of nanotechnology in diagnostic imaging and drug delivery, the engineered nanocarriers still exhibit substantial lack of hemocompatibility. Thus, when injected systemically, nanoparticles are avidly recognized by blood leukocytes and platelets, but the mechanisms of immune recognition are
A K Giri
Mutation research, 386(1), 25-38 (1997-03-01)
Epichlorohydrin (ECH) is one of the more commercially important aliphatic epoxides used extensively as an industrial intermediate, a laboratory reagent, and as an insecticide. It is a volatile, colourless liquid with an ethereal odour. It behaves as an alkylating agent.
Guang-Peng Wu et al.
Journal of the American Chemical Society, 133(38), 15191-15199 (2011-08-23)
Selective transformations of carbon dioxide and epoxides into biodegradable polycarbonates by the alternating copolymerization of the two monomers represent some of the most well-studied and innovative technologies for potential large-scale utilization of carbon dioxide in chemical synthesis. For the most
Jingyun Wang et al.
Biomacromolecules, 13(2), 559-564 (2012-01-20)
The stability of a paper-immobilized antibody was investigated over a range of temperatures (40-140 °C) and relative humidities (RH, 30-90%) using both unmodified filter paper and the same paper impregnated with polyamide-epichlorohydrin (PAE) as supports. Antibody stability decreased with increasing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service