Skip to Content
Merck
All Photos(3)

Key Documents

480746

Sigma-Aldrich

Bismuth(III) citrate

99.99% trace metals basis, −325 mesh

Synonym(s):

Bismuth citrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$66.90
500 G
$224.00

$66.90


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$66.90
500 G
$224.00

About This Item

Linear Formula:
[O2CCH2C(OH)(CO2)CH2CO2]Bi
CAS Number:
Molecular Weight:
398.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$66.90


Estimated to ship onMay 26, 2025


Request a Bulk Order

Assay

99.99% trace metals basis

reaction suitability

core: bismuth
reagent type: catalyst

particle size

−325 mesh

mp

300 °C (dec.) (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

OC12CC(=O)O[Bi](OC(=O)C1)OC2=O

InChI

1S/C6H8O7.Bi/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+3/p-3

InChI key

ANERHPOLUMFRDC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Bismuth(III)citrate is a main-group metal complex of bismuth(III) with three citrateligands. The complex has been used as a bismuth (III) source for Bi(III)radiolabeled compounds.[1]

Application

Bismuth(III) citrate has been used as a sorbent in spin columns to develop a novel stationary phase for the selective and efficient isolation of phenolic compounds from natural sources via solid phase extraction (SPE) method.[2] It can be used to synthesize bismuth subcarbonate ((BiO)2CO3) nanotubes that are reported to show antibacterial property against Helicobacter pylori.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christos Kokkinos et al.
Analytica chimica acta, 886, 29-36 (2015-09-01)
A novel immunosensor based on graphite screen-printed electrodes (SPEs) modified with bismuth citrate was developed for the voltammetric determination of C-reactive protein (CRP) in human serum using quantum dots (QDs) labels. The sandwich-type immunoassay involved physisorption of CRP capture antibody
An octadentate bis(semicarbazone) macrocycle: a potential chelator for lead and bismuth radiopharmaceuticals
Jaclyn L. Lange, et al.
Dalton Transactions, 49, 14962-14974 (2020)
Rong Chen et al.
Chemical communications (Cambridge, England), (21)(21), 2265-2267 (2006-05-24)
Bismuth subcarbonate ((BiO)2CO3) nanotubes with uniform diameters of about 3-5 nm were fabricated from bismuth citrate; these nanotubes exhibit antibacterial properties against Helicobacter pylori (50% inhibition at 10 microg mL(-1)), a bacterium causing peptic ulcers and gastritis.
Carlos Ayala-Torres et al.
Microbiological research, 176, 1-6 (2015-06-14)
Mycobacterium smegmatis Pma1 is the orthologue of M. tuberculosis P-type ATPase cation transporter CtpF, which is activated under stress conditions, such as hypoxia, starvation and response to antituberculous and toxic substances. The function of Pma1 in the mycobacterial processes across
Shah Hussain et al.
Journal of pharmaceutical and biomedical analysis, 84, 148-158 (2013-07-09)
Galloyl- and caffeoylquinic acids are among the most important pharmacological active groups of natural compounds. This study describes a pre-step in isolation of some selected representatives of these groups from biological samples. A selective solid-phase extraction (SPE) method for these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service