Skip to Content
Merck
All Photos(2)

Key Documents

478407

Sigma-Aldrich

Methyl isocyanatoformate

90%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$388.00

$388.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$388.00

About This Item

Linear Formula:
OCNCO2CH3
CAS Number:
Molecular Weight:
101.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$388.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

90%

refractive index

n20/D 1.413 (lit.)

bp

97 °C (lit.)

density

1.251 g/mL at 25 °C (lit.)

functional group

amine
isocyanate

storage temp.

−20°C

SMILES string

COC(=O)N=C=O

InChI

1S/C3H3NO3/c1-7-3(6)4-2-5/h1H3

InChI key

YOVQDZPAVARTTR-UHFFFAOYSA-N

General description

Methyl isocyanatoformate, also known as methyl carbonisocyanatidate, is an organic building block containing an isocyanate group.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service