Skip to Content
Merck
All Photos(1)

Key Documents

477850

Sigma-Aldrich

PAMAM-OH dendrimer, generation 4 solution

10 wt. % in methanol

Synonym(s):

hydroxyl terminated Polyamidoamine

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$442.00
10 G
$1,400.00

$442.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
$442.00
10 G
$1,400.00

About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=4);dendri PAMAM(NHCH2CH2OH)64
Molecular Weight:
14277.19
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$442.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

amidoethanol surface groups

Quality Level

form

solution

mol wt

generation 4

feature

core type ethylenediamine core (2-carbon core)

concentration

10 wt. % in methanol

no. Surface Groups

64

refractive index

n20/D 1.359

bp

65 °C

density

0.947 g/mL at 25 °C

storage temp.

2-8°C

Application


  • Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2.: This study explores the use of titanium dioxide (TiO2) for the photocatalytic oxidation of 2-chloroethyl ethyl sulfide, a compound related to diethyl disulfide, focusing on its potential for environmental decontamination applications (Martyanov et al., 2003).



Physical form

Contains 64 surface hydroxyl groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service