Skip to Content
Merck
All Photos(1)

Key Documents

477257

Sigma-Aldrich

1-(tert-Butyldimethylsilyloxy)-2-propanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2OCH2COCH3
CAS Number:
Molecular Weight:
188.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.426 (lit.)

bp

35-38 °C/0.6 mmHg (lit.)

density

0.976 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CO[Si](C)(C)C(C)(C)C

InChI

1S/C9H20O2Si/c1-8(10)7-11-12(5,6)9(2,3)4/h7H2,1-6H3

InChI key

LHYDYTSJUGPFLA-UHFFFAOYSA-N

General description

1-(tert-Butyldimethylsilyloxy)-2-propanone is also known as 1-{[dimethyl(2-methyl-2-propanyl)silyl]oxy}acetone. Its enthalpy of vaporization at boiling point has been reported.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 489-489 (2008)
Byung Gon Jo et al.
Molecular medicine (Cambridge, Mass.), 26(1), 119-119 (2020-12-05)
Increasing number of studies provide evidence that the vagus nerve stimulation (VNS) dampens inflammation in peripheral visceral organs. However, the effects of afferent fibers of the vagus nerve (AFVN) on anti-inflammation have not been clearly defined. Here, we investigate whether
Luca Bragina et al.
Frontiers in neuroanatomy, 12, 74-74 (2018-09-27)
Neurofilaments (NFs) are neuron-specific heteropolymers that have long been considered as structural proteins. However, it has recently been documented that they may play a functional role at synapses. Indeed, the four NF subunits-NFL, NFM, NFH and α-internexin-are integral components of
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service