Skip to Content
Merck
All Photos(1)

Key Documents

476951

Sigma-Aldrich

(2S,3S)-1,2-Epoxy-3-(Boc-amino)-4-phenylbutane

99%

Synonym(s):

tert-Butyl [S-(R*,R*)]-(−)-(1-oxiranyl-2-phenylethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO3
CAS Number:
Molecular Weight:
263.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]23/D −7°, c = 0.6 in methanol

mp

125-127 °C (lit.)

functional group

amine
ether
phenyl

SMILES string

[H][C@@]1(CO1)[C@H](Cc2ccccc2)NC(=O)OC(C)(C)C

InChI

1S/C15H21NO3/c1-15(2,3)19-14(17)16-12(13-10-18-13)9-11-7-5-4-6-8-11/h4-8,12-13H,9-10H2,1-3H3,(H,16,17)/t12-,13+/m0/s1

InChI key

NVPOUMXZERMIJK-QWHCGFSZSA-N

Application

(2S,3S)-1,2-Epoxy-3-(Boc-amino)-4-phenylbutane can be used as a reactant to prepare:
  • Hydroxyethyl urea peptidomimetics as potent γ-secretase inhibitors.[1][2][3]
  • Arylsufonamide derivatives.[4]
  • Peptidomimetic β-secretase inhibitors incorporating hydroxyethylamine isosteres.[5]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure-based design of novel HIV-1 protease inhibitors to combat drug resistance
Ghosh AK, et al.
Journal of medicinal chemistry, 49(17), 5252-5261 (2006)
Discovery of HIV-1 protease inhibitors with picomolar affinities incorporating N-aryl-oxazolidinone-5-carboxamides as novel P2 ligands
Ali Akbar, et al.
Journal of medicinal chemistry, 49(25), 7342-7356 (2006)
Arun K Ghosh et al.
Journal of medicinal chemistry, 49(17), 5252-5261 (2006-08-18)
Structure-based design and synthesis of novel HIV protease inhibitors are described. The inhibitors are designed specifically to interact with the backbone of HIV protease active site to combat drug resistance. Inhibitor 3 has exhibited exceedingly potent enzyme inhibitory and antiviral
William P Esler et al.
Bioorganic & medicinal chemistry letters, 14(8), 1935-1938 (2004-03-31)
(Hydroxyethyl)urea peptidomimetics are potent inhibitors of gamma-secretase that are accessible in a few synthetic steps. Systematic alteration of P2-P4' revealed that the corresponding S2-S4' active site pockets accommodate a variety of substituents, consistent with the fact that this protease cleaves
Probing pockets S2-S4? of the ?-secretase active site with (hydroxyethyl) urea peptidomimetics
Esler WP, et al.
Bioorganic & Medicinal Chemistry Letters, 14(8), 1935-1938 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service