Skip to Content
Merck
All Photos(1)

Key Documents

47632

Sigma-Aldrich

Fmoc-Arg(Mtr)-OH

≥98.0% (TLC)

Synonym(s):

Nα-Fmoc-Nω-(4-methoxy-2,3,6-trimethylbenzenesulfonyl)-L-arginine, Nα-Fmoc-Nω-MTR-L-arginine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H36N4O7S
CAS Number:
Molecular Weight:
608.71
Beilstein:
5206491
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥98.0% (TLC)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

COc1cc(C)c(c(C)c1C)S(=O)(=O)NC(=N)NCCC[C@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O

InChI

1S/C31H36N4O7S/c1-18-16-27(41-4)19(2)20(3)28(18)43(39,40)35-30(32)33-15-9-14-26(29(36)37)34-31(38)42-17-25-23-12-7-5-10-21(23)22-11-6-8-13-24(22)25/h5-8,10-13,16,25-26H,9,14-15,17H2,1-4H3,(H,34,38)(H,36,37)(H3,32,33,35)/t26-/m0/s1

InChI key

LKGHIEITYHYVED-SANMLTNESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Protected arginine derivative used in solid phase peptide synthesis. The MTR-guanidine protection of arg is more acid-sensitive than the tosyl group

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E. Atherton et al.
Journal of the Chemical Society. Perkin Transactions 1, 2065-2065 (1985)
M P Williamson et al.
European journal of biochemistry, 158(3), 527-536 (1986-08-01)
The peptide alpha Ahx-Met-Ala-Asp-Pro-Asn-Arg-Phe-Arg-Gly-Lys-Asp-Leu-Pro-Val-Leu- Asp-Gln-Leu-Thr-Asp-Pro-Pro-alpha Ahx (epsilon Ahx = 6-aminohexanoyl), the antigenic sequence 11-32 from Herpes simplex virus glycoprotein D-1, has been synthesised. Its 1H-NMR spectrum has been assigned by a combination of two-dimensional techniques in H2O and 2H2O. Its
M. Fujino et al.
Chemical & Pharmaceutical Bulletin, 29, 2825-2825 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service