Skip to Content
Merck
All Photos(1)

Documents

476153

Sigma-Aldrich

Diisobutyl fumarate

98%

Synonym(s):

Diisobutyl fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2O2CCH=CHCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
228.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

refractive index

n20/D 1.443 (lit.)

bp

249 °C (lit.)

mp

8 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)\C=C\C(=O)OCC(C)C

InChI

1S/C12H20O4/c1-9(2)7-15-11(13)5-6-12(14)16-8-10(3)4/h5-6,9-10H,7-8H2,1-4H3/b6-5+

InChI key

RSRICHZMFPHXLE-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balyn W Zaro et al.
Journal of immunology (Baltimore, Md. : 1950), 202(9), 2737-2746 (2019-03-20)
Dimethyl fumarate (DMF) is a prescribed treatment for multiple sclerosis and has also been used to treat psoriasis. The electrophilicity of DMF suggests that its immunosuppressive activity is related to the covalent modification of cysteine residues in the human proteome.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service