Skip to Content
Merck
All Photos(3)

Key Documents

475890

Sigma-Aldrich

2,6-Dichlorophenyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$99.90

$99.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$99.90

About This Item

Linear Formula:
Cl2C6H3NCS
CAS Number:
Molecular Weight:
204.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$99.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

38-42 °C (lit.)

functional group

chloro
isothiocyanate

storage temp.

2-8°C

SMILES string

Clc1cccc(Cl)c1N=C=S

InChI

1S/C7H3Cl2NS/c8-5-2-1-3-6(9)7(5)10-4-11/h1-3H

InChI key

SUCGVQHNGIQXGD-UHFFFAOYSA-N

General description

2,6-Dichlorophenyl isothiocyanate is an isothiocyanate derivative. It has been synthesized by using 2,6-dichlorobenzaldehyde oxime as a starting reagent.[1]

Application

2,6-Dichlorophenyl isothiocyanate may be used as a precursor to synthesize 1-(2-aminoethyl)-2-cyano-3-(2,6-dichlorophenyl)guanidine.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Tilley et al.
Journal of medicinal chemistry, 23(12), 1438-1439 (1980-12-01)
Starting with 2,6-dichlorophenyl isothiocyanate, 1-(2-aminoethyl)-2-cyano-3-(2,6-dichlorophenyl)guanidine (2) was prepared in three steps. In contrast to the corresponding thiourea 1, this compound was essentially inactive as an antihypertensive agent.
Birsa ML, et al.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 222-222 (2014)
Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service