Skip to Content
Merck
All Photos(1)

Key Documents

474533

Sigma-Aldrich

(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$52.80

$52.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$52.80

About This Item

Linear Formula:
CH3CH(OC6H4OH)CO2H
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

$52.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

optical activity

[α]20/D +58°, c = 1 in methanol

mp

145-148 °C (lit.)

functional group

carboxylic acid

SMILES string

C[C@@H](Oc1ccc(O)cc1)C(O)=O

InChI

1S/C9H10O4/c1-6(9(11)12)13-8-4-2-7(10)3-5-8/h2-6,10H,1H3,(H,11,12)/t6-/m1/s1

InChI key

AQIHDXGKQHFBNW-ZCFIWIBFSA-N

Application

(R)-2-(4-Hydroxyphenoxy)propionic acid can be used as a ligand to prepare:
  • Triorganotin(IV) complexes of biological importance.[1]
  • Co(II) based chiral coordination polymers having catalytic activity towards aldol reaction.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses, characterizations, crystal structures, and in vitro antitumor activities of chiral triorganotin (IV) complexes containing (S)-(+)-2-(4-isobutyl-phenyl) propionic and (R)-(+)-2-(4-hydroxyphenoxy) propionic acid ligands
Shi Y, et al.
Journal of Coordination Chemistry, 65(23), 4125-4136 (2012)
Syntheses, structures and catalytic properties of chiral Co (II) coordination polymers based on (R)-4-(4-(1-carboxyethoxy) phenoxy) benzoic acid
Xu X, et al.
Polyhedron, 112(23), 61-66 (2016)
Rosalía López-Ruiz et al.
Ecotoxicology and environmental safety, 157, 285-291 (2018-04-09)
A degradation study of quizalofop-p and its commercial products (quizalofop-p-ethyl, quizalofop-p-tefuryl and propaquizafop) in water samples has been performed using ultra high-performance liquid chromatography coupled to Orbitrap mass spectrometry (UHPLC-Orbitrap-MS). CHHQ (dihydroxychloroquinoxalin), CHQ (6-chloroquinoxalin-2-ol) and PPA ((R)-2-(4-hydroxyphenoxy)propionicacid) were the main

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service