Skip to Content
Merck
All Photos(1)

Key Documents

473464

Sigma-Aldrich

tert-Butylchlorodimethylsilane solution

50 wt. % in toluene

Synonym(s):

tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMSCl solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$190.00
100 G
$242.00

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$190.00
100 G
$242.00

About This Item

Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72
Beilstein:
505999
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

concentration

50 wt. % in toluene

refractive index

n20/D 1.46

bp

113 °C

density

0.87 g/mL at 20 °C (lit.)
0.866 g/mL at 25 °C

SMILES string

CC(C)(C)[Si](C)(C)Cl

InChI

1S/C6H15ClSi/c1-6(2,3)8(4,5)7/h1-5H3

InChI key

BCNZYOJHNLTNEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1A - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akshanth R Polepally et al.
Journal of clinical pharmacology, 55(10), 1101-1108 (2015-04-24)
A classic 2-period crossover bioavailability study was conducted to evaluate the relative and absolute bioavailability of immediate-release (IR) and extended-release (XR) lamotrigine formulations under steady-state conditions in elderly patients with epilepsy. On treatment days, each subject's morning dose (IR or
P Huttenloch et al.
Environmental science & technology, 35(21), 4260-4264 (2001-11-23)
The efficacy of the surface modification of natural diatomite and zeolite material by chlorosilanes is demonstrated. Chlorosilanes used were trimethylchlorosilane (TMSCI), tert-butyldimethylchlorosilane (TBDMSCI), dimethyloctadecylchlorosilane (DMODSCI), and diphenyldichlorosilane (DPDSCI) possessing different headgroups and chemical properties. Silanol groups of the diatomite and
Some observations on the t-butyldimethylchlorosilane derivatization reaction.
K Bricknell et al.
Biochemical medicine, 23(1), 119-121 (1980-02-01)
Hiroaki Wakimoto et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(11), 2898-2909 (2014-04-10)
Isocitrate dehydrogenase (IDH) gene mutations occur in low-grade and high-grade gliomas. We sought to identify the genetic basis of malignant phenotype heterogeneity in IDH-mutant gliomas. We prospectively implanted tumor specimens from 20 consecutive IDH1-mutant glioma resections into mouse brains and
C Tellis et al.
Journal of biochemistry, 119(4), 823-827 (1996-04-01)
1-O-Hexadecyl-2-acetyl-sn-glycerol, the immediate precursor of platelet- activating factor (PAF) in its de novo formation, was detected in the protozoon Tetrahymena pyriformis. It was purified from the total lipid extract by TLC, after successive developments in two different solvent systems. Characterization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service