Skip to Content
Merck
All Photos(1)

Documents

47307

Sigma-Aldrich

Fmoc-4-Abz-OH

≥96.0% (TLC)

Synonym(s):

4-(Fmoc-amino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17NO4
CAS Number:
Molecular Weight:
359.37
Beilstein:
7722986
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥96.0% (TLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(NC(=O)OCC2c3ccccc3-c4ccccc24)cc1

InChI

1S/C22H17NO4/c24-21(25)14-9-11-15(12-10-14)23-22(26)27-13-20-18-7-3-1-5-16(18)17-6-2-4-8-19(17)20/h1-12,20H,13H2,(H,23,26)(H,24,25)

InChI key

VGSYYBSAOANSLR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Li Ho et al.
Journal of biomedicine & biotechnology, 2011, 101497-101497 (2011-06-11)
Molecular imaging with promise of personalized medicine can provide patient-specific information noninvasively, thus enabling treatment to be tailored to the specific biological attributes of both the disease and the patient. This study was to investigate the characterization of DO3A-CH(2)CO-G-4-aminobenzoyl-Q-W-A-V-G-H-L-M-NH(2) (AMBA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service