Skip to Content
Merck
All Photos(1)

Key Documents

471178

Sigma-Aldrich

Benzophenone-d10

99 atom % D

Synonym(s):

Di(phenyl-2,3,4,5,6)-methanone-d10

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$929.00
5 G
$2,220.00

$929.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$929.00
5 G
$2,220.00

About This Item

Linear Formula:
(C6D5)2CO
CAS Number:
Molecular Weight:
192.28
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$929.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % D

form

solid

bp

305 °C (lit.)

mp

47-51 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(=O)c2c([2H])c([2H])c([2H])c([2H])c2[2H]

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

RWCCWEUUXYIKHB-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henry Zúñiga-Benítez et al.
Journal of environmental management, 217, 929-938 (2018-04-22)
Benzophenones (BPs) family is one of the most frequently used groups of UV-filters. However, it has been reported by different authors that this kind of chemical compounds could be associated with some endocrine disrupting activity, genotoxicity and reproductive toxicity. In
Henry Zúñiga-Benítez et al.
Journal of environmental management, 167, 246-258 (2015-12-22)
Benzophenone-3 (BP3) is one of the most used UV filters. Its disruptive effect on the endocrine system of different living beings has been demonstrated by several research groups. Present work addresses on a photocatalytic degradation of BP3 using particles of
Żaneta Broniowska et al.
Neurotoxicity research, 36(1), 39-48 (2019-04-23)
Benzophenones, frequently used as UV chemical filters, are absorbed through the skin and can exert systemic adverse effects. So far, most of the data are related to their action on sex hormone receptors whereas potential neurotoxic effect is expected mainly
Ming Zheng et al.
Journal of hazardous materials, 402, 123484-123484 (2020-07-31)
Trace organic contaminants (TOrCs) in waste active sludge (WAS) have caused many concerns due to their recalcitrance and detriment to the performance of anaerobic digestion (AD). In this study, UV (2 h) combined with calcium peroxide (CaO2, 0.1 g g-1-VSS
Víctor Vállez-Gomis et al.
Journal of chromatography. A, 1624, 461229-461229 (2020-06-17)
This work describes a sensitive and rapid analytical method for trace determination of polycyclic aromatic hydrocarbons (PAHs) in cosmetic samples. The proposed method is based on stir bar sorptive-dispersive microextraction (SBSDME). A magnetic composite made of CoFe2O4 magnetic nanoparticles embedded

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service