Skip to Content
Merck
All Photos(1)

Key Documents

470163

Sigma-Aldrich

3-(Chloromethyl)pentane

95%

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$156.00

$156.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$156.00

About This Item

Linear Formula:
(C2H5)2CHCH2Cl
CAS Number:
Molecular Weight:
120.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$156.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Assay

95%

refractive index

n20/D 1.423 (lit.)

bp

126-128 °C/753 mmHg (lit.)

density

0.891 g/mL at 25 °C (lit.)

functional group

alkyl halide
chloro

SMILES string

CCC(CC)CCl

InChI

1S/C6H13Cl/c1-3-6(4-2)5-7/h6H,3-5H2,1-2H3

InChI key

PQHNRODYYLFLRE-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service