Skip to Content
Merck
All Photos(1)

Key Documents

469424

Sigma-Aldrich

Methyl (2R)-glycidate

optical purity ee: 94% (GLC), 97%

Synonym(s):

Methyl (R)-oxiranecarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$384.00
25 G
$1,310.00

$384.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$384.00
25 G
$1,310.00

About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$384.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

97%

optical activity

[α]20/D +32°, c = 1 in chloroform

optical purity

ee: 94% (GLC)

refractive index

n20/D 1.42 (lit.)

density

1.166 g/mL at 25 °C (lit.)

functional group

ester
ether

shipped in

wet ice

storage temp.

2-8°C

SMILES string

COC(=O)[C@H]1CO1

InChI

1S/C4H6O3/c1-6-4(5)3-2-7-3/h3H,2H2,1H3/t3-/m1/s1

InChI key

YKNYRRVISWJDSR-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

162.0 °F - closed cup

Flash Point(C)

72.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service