Skip to Content
Merck
All Photos(1)

Key Documents

468266

Sigma-Aldrich

Poly(ethylene glycol) 2,4,6-tris(1-phenylethyl)phenyl ether methacrylate solution

average Mn ~1,600, 60 wt. % in methacrylic acid/water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2(CH2CH2O)nC6H2[CH(CH3)C6H5)]3
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~1,600

contains

150-250 ppm MEHQ as stabilizer
20% water

concentration

60 wt. % in methacrylic acid/water

refractive index

n20/D 1.462

viscosity

150-175 cP(lit.)

density

1.09 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OCCO.CC(=C)C(O)=O.CC(c1ccccc1)c2cc(C(C)c3ccccc3)c(O)c(c2)C(C)c4ccccc4

Application

Copolymerizable surfactant and associative thickener in latex polymers.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service