Skip to Content
Merck
All Photos(1)

Key Documents

468207

Sigma-Aldrich

Iron(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate)

Synonym(s):

Fe(TMHD)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe[(CH3)3CCOCHCOC(CH3)3]
CAS Number:
Molecular Weight:
605.65
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: iron

mp

179-185 °C (lit.)

SMILES string

CC(C)(C)C(=O)\C=C(\O[Fe](O\C(=C\C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/3C11H20O2.Fe/c3*1-10(2,3)8(12)7-9(13)11(4,5)6;/h3*7,12H,1-6H3;/q;;;+3/p-3/b2*8-7+;8-7-;

InChI key

NXBJVTQWYMSPSJ-GECNZSFWSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Precursor for the formation of BaFe12O19 thin films on Al2O3 (0001) substrates in preferential orientation with the c-axis perpendicular to the substrate. Precursor for aerosol-assisted CVD of mixed-conducting ceramic films of Sr-Co-Fe perovskite phases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calvin D Pham et al.
ACS applied materials & interfaces, 9(42), 36980-36988 (2017-09-20)
A radical-enhanced atomic layer deposition (RE-ALD) process was developed for growing ferrimagnetic CoFe

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service