Skip to Content
Merck
All Photos(1)

Key Documents

468010

Sigma-Aldrich

3-Hydroxybenzoic hydrazide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CONHNH2
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Assay

98%

mp

158-161 °C (lit.)

SMILES string

NNC(=O)c1cccc(O)c1

InChI

1S/C7H8N2O2/c8-9-7(11)5-2-1-3-6(10)4-5/h1-4,10H,8H2,(H,9,11)

InChI key

OACGSLLKFCMXSX-UHFFFAOYSA-N

Application

3-Hydroxybenzoic hydrazide may be used in the following studies:
  • As starting reagent in the preparation of hydrazide intermediates, required for the synthesis of the brevenal analogs.[1]
  • Synthesis of N′-[4-(dimethylamino)benzylidene]-3-hydroxybenzohydrazide.[2]
  • Synthesis of N′-(5-bromo-2-hydroxybenzylidene)-3-hydroxybenzohydrazide.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N'-(5-Bromo-2-hydroxybenzylidene)-3-hydroxybenzohydrazide.
Nie Y.
Acta Crystallographica Section E, Structure Reports Online, 64(2), o514-o514 (2008)
Allan Goodman et al.
Marine drugs, 12(4), 1839-1858 (2014-04-02)
Brevenal is a ladder frame polyether produced by the dinoflagellate Karenia brevis. This organism is also responsible for the production of the neurotoxic compounds known as brevetoxins. Ingestion or inhalation of the brevetoxins leads to adverse effects such as gastrointestinal
N'-[4-(Dimethylamino) benzylidene]-3-hydroxybenzohydrazide.
Nie Y.
Acta Crystallographica Section E, Structure Reports Online, 64(2), 471-471 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service