Skip to Content
Merck
All Photos(2)

Documents

467383

Sigma-Aldrich

Cystamine sulfate hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S2(CH2CH2NH2)2·H2SO4·xH2O
CAS Number:
Molecular Weight:
250.36 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

278 °C (dec.) (lit.)

SMILES string

O.OS(O)(=O)=O.NCCSSCCN

InChI

1S/C4H12N2S2.H2O4S.H2O/c5-1-3-7-8-4-2-6;1-5(2,3)4;/h1-6H2;(H2,1,2,3,4);1H2

InChI key

AARGHWGZRNMDIP-UHFFFAOYSA-N

Related Categories

General description

Cystamine has radiation-protective propetries and can be used to prevent normal tissues from radiation damage in clinical radiation therapy.

Application

Cystamine sulfate hydrate has been used for attaching PPO (polyphenol) on to gold nanoparticles while developing biosensor for the determination of sulfite in white and red wine samples. It may be used as starting reagent in the synthesis of cystamine bis(5-aminotetrazolate).

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal Structures of New Ammonium 5-Aminotetrazolates.
Lampl M, et al.
Crystals, 4(4), 439-=49-439-=49 (2014)
Elen Romão Sartori et al.
Talanta, 87, 235-242 (2011-11-22)
The modification of a glassy carbon electrode with multi-walled carbon nanotubes and gold nanoparticles within a poly(allylamine hydrochloride) film for the development of a biosensor is proposed. This approach provides an efficient method used to immobilize polyphenol oxidase (PPO) obtained
Ridthee Meesat et al.
Radiation research, 177(6), 813-826 (2012-04-06)
Cystamine, an organic disulfide (RSSR), is among the best of the known radiation-protective compounds and has been used to protect normal tissues in clinical radiation therapy. Recently, it has also proved to be beneficial in the treatment of disorders of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service