Skip to Content
Merck
All Photos(3)

Key Documents

467227

Sigma-Aldrich

2-Ethynylanisole

97%

Synonym(s):

1-Ethynyl-2-methoxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$140.00
5 G
$585.00

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$140.00
5 G
$585.00

About This Item

Linear Formula:
HC≡CC6H4OCH3
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$140.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.5720 (lit.)

bp

197 °C (lit.)

density

1.022 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1C#C

InChI

1S/C9H8O/c1-3-8-6-4-5-7-9(8)10-2/h1,4-7H,2H3

InChI key

UFOVULIWACVAAC-UHFFFAOYSA-N

General description

2-Ethynylanisole is an acetylene derivative.[1] It has been prepared by reacting 2-iodoanisole with trimethylsilylacetylene followed by the deprotection of trimethylsilyl group.[2][3]

Application

2-Ethynylanisole may be used in the synthesis of 1,4-bis(2-methoxyphenyl)-2-methylbenzene.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

196.0 °F - closed cup

Flash Point(C)

91.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Π-conjugated polymers via regioregular organometallic polymers 2. Transformation of titanacyclopentadiene-containing polymer into poly (p-phenylene) derivative.
Kino THN and Tomita I.
Applied Organometallic Chemistry, 24(8), 558-562 (2010)
The acid-catalysed synthesis of 7-azaindoles from 3-alkynyl-2-aminopyridines and their antimicrobial activity.
Leboho TC, et al.
Organic & Biomolecular Chemistry, 12(2), 307-315 (2014)
T Iijima et al.
Chemical & pharmaceutical bulletin, 47(3), 398-404 (1999-04-23)
Synthesis and biological evaluation of the first dicarba-closo-dodecaborane (carborane) derivatives of retinoids are described. Their retinoidal activity were examined in terms of the differentiation-inducing ability toward human promyelocytic leukemia HL-60 cells. High retinoidal activity (agonist or antagonist for retinoic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service