Skip to Content
Merck
All Photos(1)

Key Documents

466301

Sigma-Aldrich

Nickel(II) iodide

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Nickel diiodide, Nickel iodide, Nickelous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiI2
CAS Number:
Molecular Weight:
312.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder
solubility:
H2O: slightly soluble(lit.)
alcohols: slightly soluble(lit.)

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

797 °C (lit.)

solubility

H2O: slightly soluble(lit.)
alcohols: slightly soluble(lit.)

density

5.83 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

I[Ni]I

InChI

1S/2HI.Ni/h2*1H;/q;;+2/p-2

InChI key

BFSQJYRFLQUZKX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Strongly accelerates Sml2 mediated homocoupling of imines into vicinal diamines and heterocoupling reactions between imines and ketones.[1]

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adél Anna Ádám et al.
Nanomaterials (Basel, Switzerland), 10(4) (2020-04-02)
In the experimental work leading to this contribution, the parameters of the ultrasound treatment (temperature, output power, emission periodicity) were varied to learn about the effects of the sonication on the crystallization of Ni nanoparticles during the hydrazine reduction technique.
Machrouhi, F. Namy, J-L.
Tetrahedron Letters, 40, 1315-1315 (1999)

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service