Skip to Content
Merck
All Photos(3)

Key Documents

465577

Sigma-Aldrich

Silicon 2,9,16,23-tetra-tert-butyl-29H,31H-phthalocyanine dihydroxide

Dye content 80 %

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$429.00

$429.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$429.00

About This Item

Empirical Formula (Hill Notation):
C48H50N8O2Si
CAS Number:
Molecular Weight:
799.05
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$429.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

composition

Dye content, 80%

mp

>300 °C (lit.)

λmax

678 nm

SMILES string

CC(C)(C)c1ccc2c3nc(nc4n5c(nc6nc(nc7n(c(n3)c8cc(ccc78)C(C)(C)C)[Si]5(O)O)c9cc(ccc69)C(C)(C)C)c%10cc(ccc4%10)C(C)(C)C)c2c1

InChI

1S/C48H50N8O2Si/c1-45(2,3)25-13-17-29-33(21-25)39-49-37(29)51-43-36-24-28(48(10,11)12)16-20-32(36)42-54-40-34-22-26(46(4,5)6)14-18-30(34)38(50-40)52-44-35-23-27(47(7,8)9)15-19-31(35)41(53-39)55(44)59(57,58)56(42)43/h13-24,57-58H,1-12H3/b51-37-,51-43-,52-38-,52-44-,53-39-,53-41-,54-40-,54-42-

InChI key

YTJFELPKNFDSGI-ONAHWTHPSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service